4-Amino-3-hydroxy-1-naphthalenesulfonic acid for analysis of phosphate, Supelco®

Supplier: MilliporeSigma

1.00099.0025
CA1.00099.0025CS 45839 CAD
CA1.00099.0025
4-Amino-3-hydroxy-1-naphthalenesulfonic acid for analysis of phosphate, Supelco®
4-Amino-3-hydroxy-1-naphthalenesulphonic acid
Formula: C₁₀H₉NO₄S
Boiling Pt: 466 °C (1013 hPa)
Melting Pt: ∼285 °C
Storage Temperature: Ambient
MDL Number: MFCD00004019
CAS Number: 116-63-2
EINECS: 204-147-3
Merck Index: 13,00453

Order Now

Specification Test Results

Solubility (20 °C) practically insoluble
Molar Mass 239.24 g/mol
Bulk Density 280 kg/m3


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR