Toluene ≥99.5% (by GC), GR ACS, Supelco®

Supplier: MilliporeSigma
Danger

TX0735-20 TX0735-3 TX0735-5 TX0735-6 TX0735T-4
CATX0735-20EA 3396 CAD
CATX0735-20 CATX0735-3 CATX0735-5 CATX0735-6 CA-TX0735T-4
Toluene ≥99.5% (by GC), GR ACS, Supelco®
Toluene

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs.

Formula: C₆H₅CH₃
Boiling Pt: 110…111 °C (lit.)
Melting Pt: –93 °C (lit.)
Density: 0.865 g/ml (25°C) (lit.)
Flash Pt: 4 °C
Storage Temperature: Ambient
MDL Number: MFCD00008512
CAS Number: 108-88-3
EINECS: 203-625-9
UN: 1294
ADR: 3,II
REACH: 05-2114615130-69
Merck Index: 13,09607
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44efa31292a7dce6d9e60bcbaec208eb42d34e32&vid=73ada9ad1e894808b6c193cabc4babcfaf320a25&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) 99.5% min
Color (APHA) 10 max
Form Clear liquid
Infrared spectrum Conforms to standard
Residue after evaporation 0.001% max
Substances darkened by sulfuric acid To pass test
Sulfur compounds (as S) 0.003% max
Water (H2O) 0.03% max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR