Sodium acetate, anhydrous ≥99.0%, OmniPur®, Millipore®

Supplier: MilliporeSigma

7510-500GM 7560-5KG
CA-EM7510EA 121.2 CAD
CA-EM7510 CA-EM7560
Sodium acetate, anhydrous ≥99.0%, OmniPur®, Millipore®
Sodium acetate

DNase-, RNase, protease-, and phosphatase-free.

Formula: H₃CCOONa
Boiling Pt: 881.4 °C (1013 hPa)
Melting Pt: 324 °C
Density: 1.528 g/cm³ (20 °C)
Flash Pt: >250 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00012459
CAS Number: 127-09-3
EINECS: 204-823-8
REACH: 01-2119485123-42
Merck Index: 12,08711

Order Now

Specification Test Results

A (260 nm, 0.1 M, water) 0.01AU max
A (280 nm, 0.1 M, water) 0.01AU max
Appearance White crystals
Assay 99.0% min
DNase None detected
Heavy metals (as Pb) 0.001% max
pH (5%, water) 7.5-9.2
Phosphatase None detected
Protease None detected
RNase None detected


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR