Cyclohexane 99 GC_ASSAY_METHOD, GR ACS ACS, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Hexahydrobenzene, Hexamethylene

CX2290-1 CX2290-5 CX2290-30
CACX2290-1EA 469 CAD
CACX2290-1 CACX2290-5 CA-CX2290-30 CACX2290-3
Cyclohexane 99 GC_ASSAY_METHOD, GR ACS ACS, Supelco®
Cyclohexane

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs

Formula: C₆H₁₂
MW: 84.16 g/mol
Boiling Pt: 81 (lit.)
Melting Pt: 7 (lit.)
Density: 0.779 g/ml (25°C) (lit.)
Flash Pt: –18 °C
Storage Temperature: Ambient
MDL Number: MFCD00003814
CAS Number: 110-82-7
EINECS: 203-806-2
UN: 1145
ADR: 3 II
Merck Index: 13,02752
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a1e51ba998bef126804c3fd25b361be72cd9545&vid=d1e7f75d9f2a944eb15a7740c5254588cc9a4e67&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) 99.0% min
Appearance Clear
Color (APHA) 10 max
Form Liquid
Infrared spectrum Conforms to standard
Residue after evaporation 0.002% max
Substances darkened by sulfuric acid To pass test
Water (H2O) 0.02% max

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR