Dichloroacetic acid (DCA) 10% in toluene deblocking reagent

Supplier: MilliporeSigma

Synonyms: DCA Deblock, DCA

BI0841-4000 BI0841-NP20
CA-BI0841-4000EA 341 CAD
CA-BI0841-4000 CABI0841-NP20
Dichloroacetic acid (DCA) 10% in toluene deblocking reagent
Dichloroacetic acid (DCA)

These deblocking reagents are mainly involved in DNA synthesis.


  • Infrared spectrum conforms to the standard
  • Corrosive and toxic
  • Clear liquid form

Formula: C₂H₂Cl₂O₂
MW: 128.94 g/mol
Boiling Pt: 194
Melting Pt: -4
MDL Number: MFCD00004223
CAS Number: 79-43-6
ADR: 8 II
Merck Index: 13,03075

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR