Corning® BioCoat® Poly-D-Lysine Plates, Corning
Supplier: Corning
356509 356413 356414 356470 354509 354413 354414 354470
CA62406-281CS 1176.91 CAD
CA62406-281 CACB356413 CACB356414 CACB356470 CA-62406-279 CA-CB354413 CA-CB354414 CA-CB354470
Corning® BioCoat® Poly-D-Lysine Plates, Corning
Microplates Cell Culture Plates
Poly-D-Lysine (PDL) and Poly-L-Lysine (PLL) are synthetic compounds that enhance cell adhesion and protein absorption by altering surface charges on the culture substrate. In addition to promoting cell adhesion, poly-lysine surface treatments support neurite outgrowth and improve the survival of many central nervous system (CNS) primary cells in culture. As PDL and PLL are synthetic molecules, they do not stimulate biological activity in the cells cultured on them, and they do not introduce impurities carried by natural polymers.
  • Attachment and spreading of a variety of cell types
  • Cell
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Environmental certification