Ferroin (1,10-phenanthrolineferrous sulfate complex) solution Reag. Ph. Eur. redox indicator E0 (H2SO4 1 mol/l) = +1,06 volt, Fluka™

Supplier: Honeywell Research Chemicals

Synonyms: Tris(1,10-phenanthroline)iron (II) sulphate, Tris(1,10-phenanthroline-N1,N10)iron sulphate, Tris(1,10-phenanthroline)iron (II) sulfate, Tris(1,10-phenanthroline-N1,N10)iron sulfate

34531-100ML 34531-500ML
BJ34531-100MLEA 98.64 CAD
BJ34531-100ML BJ34531-500ML
Ferroin (1,10-phenanthrolineferrous sulfate complex) solution Reag. Ph. Eur. redox indicator E0 (H2SO4 1 mol/l) = +1,06 volt, Fluka™
Ferroin (1,10-Phenanthroline-ferrous sulphate-complex)

E0 in sulfuric acid 1 mol/l = +1,06 volt

Formula: C₃₆H₂₄FeN₆SO₄
MW: 692.54
Boiling Pt: 100 °C (1.013 hPa)
Density: 0.9999
Storage Temperature: Ambient
MDL Number: MFCD00036428
CAS Number: 14634-91-4

Order Now

Specification Test Results

Change complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR