Mercury(II) nitrate 0.01 M Hg(NO3)2 (0.02N) ≥99.5% volumetric solution, for chloride determination, Fluka™

Supplier: Honeywell Research Chemicals
Warning

Synonyms: Mercury dinitrate, Mercuric nitrate

34291-1L
BJ34291-1LEA 516.14 CAD
BJ34291-1L
Mercury(II) nitrate 0.01 M Hg(NO3)2 (0.02N) ≥99.5% volumetric solution, for chloride determination, Fluka™
Mercury (II) nitrate
Formula: HgN₂O₆
MW: 324.60 g/mol
Density: 1.005 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011038
CAS Number: 10045-94-0

Order Now

Specification Test Results

Factor 0.995 - 1.005

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR