pH Buffer Solutions, Referenced at 20 °C, Traceable to NIST, Fluka™, Honeywell Research Chemicals
Supplier: Honeywell Research Chemicals
31046-6X1L 33545-6X1L 33547-6X1L 33643-10L-VP 33643-6X1L 33643-500ML 33643-5L-VP 33648-10L-VP 33648-6X1L 33544-6X1L
BJ31046-1LCS 597.89 CAD
BJ31046-1L BJ33544-1L BJ33545-1L BJ33547-1L BJ33643-100ML BJ33643-10L-VP BJ33643-1L BJ33643-500ML BJ33643-5L-VP BJ33648-10L-VP BJ33648-1L BJ33648-5L-VP BJ33544-6X1L
pH Buffer Solutions, Referenced at 20 °C, Traceable to NIST, Fluka™, Honeywell Research Chemicals
Buffers pH Calibration Buffers
  • Citrate and phosphate buffer solutions contain fungicide
  • Traceable to SRM from NIST
New Product
Order Now
SPECIFICATIONS
Environmentally Preferable