Sodium chloride concentrate for dilution to 1L, 0.1 M NaCl (0.1N) Volumetric, Fluka™

Supplier: Honeywell Research Chemicals

Synonyms: Saline, NaCl, Sea salt, Halite

32038-1EA
BJ32038-1EAEA 125.85 CAD
BJ32038-1EA
Sodium chloride concentrate for dilution to 1L, 0.1 M NaCl (0.1N) Volumetric, Fluka™
Sodium chloride
Formula: NaCl
MW: 58.44 g/mol
Boiling Pt: 1413
Melting Pt: 801
Density: 1.00 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5
EINECS: 231-791-2
Merck Index: 14,08599

Order Now

Specification Test Results

Factor 0.998 - 1.002

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR