L(+)-Potassium sodium tartrate tetrahydrate 99.0-102.0% Reag. USP, ACS, VWR Chemicals BDH®

Supplier: VWR International

BDH9272-500G
BDH9272-500GCS 2771.56 CAD
BDH9272-500G
L(+)-Potassium sodium tartrate tetrahydrate 99.0-102.0% Reag. USP, ACS, VWR Chemicals BDH®
L(+)-Potassium sodium tartrate tetrahydrate

Colorless or white solid

Formula: KOOC(CHOH)₂COONa·4H₂O
Melting Pt: 70 to 80 °C
MDL Number: MFCD00150989
CAS Number: 6381-59-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d585522f4e3b14a7b011dc8a858f5eeca413864&vid=542d22225b124dda886e5007091b35db67214bb7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Ammonium = 0.002 %
Assay 99.0 - 102.0 %
Calcium = 0.005 %
Chloride = 0.001 %
Heavy Metals = 5 ppm
Insoluble Matter = 0.005 %
Iron = 0.001 %
pH (5%, Water) @25.0C 6.0 - 8.5
Phosphate = 0.002 %
Sulfate = 0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR