n-Pentane ≥99%, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®

Supplier: VWR International
Danger

Synonyms: Pentane

BDH83632.400
BDH83632.400CS 2434.01 CAD
BDH83632.400
n-Pentane ≥99%, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
n-Pentane
Formula: H₃C(CH₂)₃CH₃
MW: 72.15 g/mol
Boiling Pt: 35…36 °C (1013 hPa)
Melting Pt: –130 °C
Density: 0.63 g/cm³ (20 °C)
Flash Pt: –40 °C
Storage Temperature: Ambient
MDL Number: MFCD00009498
CAS Number: 109-66-0
EINECS: 203-692-4
UN: 1265
ADR: 3 II
REACH: 01-2119459286-30
Merck Index: 13,07193
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ba2c9c81a4b1f44769627a00ba49b6cc8e63132&vid=f6831eb15ab1602d8c752468dc3ee31cc4ee61c0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (by GC-FID) (as n-Pentane) Min. 99 %
Water (by KF coulometry) Max. 0.01 %
Nonvolatile Residue Max. 0.0005 %
Acidity Max. 0.0005 meq/g
Alkalinity Max. 0.0002 meq/g
Fluorescence (at emmission maximum for impurites) (254 nm) Max. 0.001 ppm
Fluorescence (at emmission maximum for impurites) (365 nm) Max. 0.001 ppm
Transmittance (200 nm) Min. 10 %
Transmittance (210 nm) Min. 40 %
Transmittance (215 nm) Min. 70 %
Transmittance (222 nm) Min. 90 %
Transmittance (240 nm) Min. 98 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR