Sodium carbonate 1 N in aqueous solution, VWR Chemicals BDH®

Supplier: VWR International

Synonyms: Calcined soda, Carbonic acid disodium salt

BDH7971-1
BDH7971-1CS 1505.5 CAD
BDH7971-1
Sodium carbonate 1 N in aqueous solution, VWR Chemicals BDH®
Sodium carbonate

Made with deionized water. Suitable as a titrant.

Formula: Na₂CO₃
MW: 105.99 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003494
CAS Number: 497-19-8
EINECS: 207-838-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a6ff30f1487cdef86add98b38bfa8eb3dac14f6&vid=c5b129b7acb5c897b9d970ea3130d61fb1934b54&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance/color Clear liquid
N.I.S.T. traceable to SRM 723 X
Normality 0.995 - 1.005 N
Shelf-life 2 years

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR