Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) solution 1 M analytical reagent pH 8.0, VWR Chemicals BDH®

Supplier: VWR International

Synonyms: 2-Amino-2-(hydroxymethyl)propane-1,3-diol(tris), Tromethamine, Tris(hydroxymethyl)methylamine, 2-Amino-2-(hydroxymethyl)propane-1,3-diol, 2-Amino-2-(hydroxymethyl)-1,3-propanediol, Trizma(R) base, TRIS base, THAM

BDH7729-1
BDH7729-1CS 3165.81 CAD
BDH7729-1
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) solution 1 M analytical reagent pH 8.0, VWR Chemicals BDH®
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol)

Made with deionized water ultra pure.

Formula: NH₂C(CH₂OH)₃
MDL Number: MFCD00004679
CAS Number: 77-86-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=11a03d1ab6b2c11b9a2c83f0a73fc4264cec0110&vid=5ce890cdf298a59a7900a07fee1c857a5e5e61e0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance/color Clear liquid
Shelf-life 18 months
pH 7.90 - 8.05


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR