Bromothymol blue sodium salt 0.04% (w/v) in aqueous solution, VWR Chemicals BDH®

Supplier: VWR International

BDH7393-2
BDH7393-2CS 1119.35 CAD
BDH7393-2
Bromothymol blue sodium salt 0.04% (w/v) in aqueous solution, VWR Chemicals BDH®
Bromothymol blue sodium salt

Made with deionized water.

Formula: C₂₇H₂₇Br₂NaO₅S
Storage Temperature: Ambient
MDL Number: MFCD00077263
CAS Number: 34722-90-2
EINECS: 252-169-7

Order Now

Specification Test Results

Appearance/color Blue liquid
Shelf-life 2 years


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR