Bromophenol blue 0.04% (w/v) in aqueous solution, VWR Chemicals BDH®

Supplier: VWR International

BDH7392-2
BDH7392-2CS 984.23 CAD
BDH7392-2
Bromophenol blue 0.04% (w/v) in aqueous solution, VWR Chemicals BDH®
Bromophenol blue

Made with deionized water.

Formula: C₁₉H₁₀Br₄O₅S
Storage Temperature: Ambient
MDL Number: MFCD00005875
CAS Number: 115-39-9
EINECS: 204-086-2

Order Now

Specification Test Results

Appearance/color Purple liquid
Shelf-life 2 years


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR