Phenolphthalein 1% in alcohol, VWR Chemicals BDH®

Supplier: VWR International
Danger

Synonyms: 3,3-Bis(4-hydroxyphenyl)isobenzofuran-1(3H)-one

BDH7344-1 BDH7344-2 BDH7344-4
BDH7344-1CS 2420.22 CAD
BDH7344-1 BDH7344-2 BDH7344-4
Phenolphthalein 1% in alcohol, VWR Chemicals BDH®
Phenolphthalein

Made with denatured ethanol.

Formula: C₂₀H₁₄O₄
MW: 318.33 g/mol
Boiling Pt: ∼78 °C (1013 hPa)
Density: ∼0.85 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005913
CAS Number: 77-09-8
EINECS: 201-004-7
UN: 1170
ADR: 3 II
Merck Index: 14,07243
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d81b539a32ba7c40a473e6afbb8e0dae525d637&vid=91bf141a7ea82baf5df563f73967884d2a4b5d79&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance/color Clear liquid

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR