Sodium hydroxide 10.0 N in aqueous solution, VWR Chemicals BDH®

Supplier: VWR International
Danger

BDH7247-1 BDH7247-10 BDH7247-4 BDH7247-5 BDH7247-7
BDH7247-1CS 972.94 CAD
BDH7247-1 BDH7247-10 BDH7247-4 BDH7247-5 BDH7247-7
Sodium hydroxide 10.0 N in aqueous solution, VWR Chemicals BDH®
Sodium hydroxide

Made with deionized water. Suitable as a titrant.


Shelf-life 2 years.

Formula: NaOH
Boiling Pt: 109 °C (1013 hPa)
Density: 1.34 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,II

Order Now

Specification Test Results

Normality 9.9 - 10.1 N
Appearance Clear liquid
Traceable to NIST SRM 84 Confirmed
Prepared to formulation on file Confirmed

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR