Sodium hydroxide 1.0 N in aqueous solution, VWR Chemicals BDH®

Supplier: VWR International
Danger

BDH7222-1 BDH7222-4 BDH7222-7
BDH7222-1CS 881.55 CAD
BDH7222-1 BDH7222-4 BDH7222-7
Sodium hydroxide 1.0 N in aqueous solution, VWR Chemicals BDH®
Sodium hydroxide

NIST traceable. Suitable as a titrant.

Formula: NaOH
Density: 1.04 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 3266
ADR: 8,III

Order Now

Specification Test Results

Appearance/color Clear liquid
Traceable to NIST SRM 84 Yes
Normality 0.995 - 1.005 N
Shelf-life 2 years
Manufacture date As reported
Expiration date As reported


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR