Iodine solution 0.1 N, VWR Chemicals BDH®

Supplier: VWR International

BDH7207-1 BDH7207-4
BDH7207-1CS 524.55 CAD
BDH7207-1 BDH7207-4
Iodine solution 0.1 N, VWR Chemicals BDH®
Iodine

Made with iodine crystals, potassium iodide and deionized water.

Formula: I₂
MDL Number: MFCD00011355
CAS Number: 7553-56-2
UN: 3495
ADR: 8,III

Order Now

Specification Test Results

Normality 0.0995 - 0.1005 N
Appearance/color Dark brown liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR