Hydrochloric acid 6.0 N, VWR Chemicals BDH®

Supplier: VWR International
Warning

Synonyms: Hydrochloric acid solution, Muriatic acid

BDH7204-1 BDH7204-4
BDH7204-1CS 2166.62 CAD
BDH7204-1 BDH7204-4
Hydrochloric acid 6.0 N, VWR Chemicals BDH®
Hydrochloric acid

Made with high purity acid and deionized water. Suitable as a titrant.

Formula: HCl
MW: 36.46
Density: 1.06 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8 II

Order Now

Specification Test Results

Appearance/color Clear liquid
Normality 5.950 - 6.050 N
Shelf-life 2 years
N.I.S.T. traceable to SRM 723 Yes


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR