Hydrobromic acid 48% ACS, VWR Chemicals BDH®

Supplier: VWR International
Danger

BDH4572-500ML
BDH4572-500MLCS 1256.85 CAD
BDH4572-500ML
Hydrobromic acid 48% ACS, VWR Chemicals BDH®
Hydrogen bromide
Formula: HBr
Density: 1.47 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011323
CAS Number: 10035-10-6
EINECS: 233-113-0
UN: 1788
ADR: 8,II

Order Now

Specification Test Results

Assay 47.0 - 49.0 %
Heavy metals (as Pb) Max. 5 ppm
Residue after ignition Max. 0.002 %
Sulphate and sulphite (as SO4) Max. 0.003 %
Cl (Chloride) Max. 0.05 %
I (Iodide) Max. 0.003 %
PO4 (Phosphate) Max. 0.001 %
Fe (Iron) Max. 1 ppm
Se (Selenium) Max. 0.01 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR