Aluminium sulfate octadecahydrate 98.0-102.0%, crystallised ACS

Supplier: VWR International
Danger

BDH4552-500G
BDH4552-500GCS 1674.71 CAD
BDH4552-500G
Aluminium sulfate octadecahydrate 98.0-102.0%, crystallised ACS
Aluminium sulphate octadecahydrate
Formula: Al₂(SO₄)₃·18H₂O
Melting Pt: 86 °C
Density: 1.69 g/cm³ (20 °C)
MDL Number: MFCD00149136
CAS Number: 7784-31-8
EINECS: 233-135-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4dace68759944aa1c6c0f56a372ea396f004e261&vid=e63de3c58650a97622a605c237aa52402ff157e4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (~14-18 H2O) 98.0 - 102.0 %
Molecules of water (x H2O) 14 - 18
Heavy metals (as Pb) Max. 0.001 %
Insoluble matter Max. 0.01 %
Cl (Chloride) Max. 0.005 %
Ca (Calcium) Max. 0.01 %
K (Potassium) Max. 0.005 %
Fe (Iron) Max. 0.002 %
Mg (Magnesium) Max. 0.002 %
Na (Sodium) Max. 0.02 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR