Water ASTM Type II, VWR Chemicals BDH®

Supplier: VWR International

Synonyms: Hydrogen oxide

BDHVBDH1168-204L BDHVBDH1168-4LP BDH1168-1LP BDHVBDH1168-5GL
BDH1168-204LEA 1432.46 CAD
BDH1168-204L BDH1168-4LP BDH1168-1LP BDH1168-5GL
Water ASTM Type II, VWR Chemicals BDH®
Water

Type II grade of reagent water is prepared by distillation using a still designed to produce a distillate having a conductivity of less than 1.0 µS/cm at 298 K (25°C).


  • Deionized
  • Used for general laboratory applications
  • Suitable for feeding ultra-pure systems, among other uses


Ion exchange, distillation, or reverse osmosis and organic adsorption may be required prior to distillation if the purity cannot be attained by single distillation.

Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100
Melting Pt: 0
Density: 1 g/cm³ (4 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Order Now

Specification Test Results

Electrical Conductivity at 25°C Less than 1.0µS/cm
Electrical Resistivity at 25°C Greater than or equal to 1.0 megohm-cm
Total Organic Carbon Less than 50 µg/L
Sodium (Na) Less than 5 µg/L
Chlorides (Cl) Less than 5 µg/L
Total Silica Less than 3 µg/L


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR