tert-Butyl methyl ether ≥99% ACS, VWR Chemicals BDH®

Supplier: VWR International
Danger


Stabilization: _

BDH1139-4LG
BDH1139-4LGCS 1138.01 CAD
BDH1139-4LG
tert-Butyl methyl ether ≥99% ACS, VWR Chemicals BDH®
tert-Butyl methyl ether

Meets ACS specifications for general use.

Formula: (CH₃)₃COCH₃
Boiling Pt: 55.2 °C (1013 hPa)
Melting Pt: –108.6 °C
Density: 0.7405 g/cm³ (20 °C)
Flash Pt: –28°C
Storage Temperature: Ambient
MDL Number: MFCD00008812
CAS Number: 1634-04-4
EINECS: 216-653-1
UN: 2398
ADR: 3,II
Merck Index: 13,06059
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1712bc714ba0a06e0de37f7cf4e5117256f84425&vid=e1b314f829136075329c50f9e5a593c19ae28b4f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear, colorless liquid
Water (K.F.) Less than 0.05%
Residue Less than 0.001% (0.0010 g/136 mL)
GC Purity Greater than 99.0%
Color (APHA) Less than 10
Peroxides (TiCl4; mg/L) Less than 1.0 mg/L as H2O2 [Typical - Not detected (less than 0.1 mg/L)]
Peroxides (TiCl4; mg/Kg) Less than 1.0 mg/Kg as H2O2 [Typical - Not detected (less than 0.1 mg/Kg)]


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR