CrossLab Graphite/Vespel Ferrules for GC
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
0100-1332 5062-3580 5062-3581 5062-3538 5181-3308 5062-3508 5181-3323 5062-3512 5062-3514 500-1004 CR213200 5181-3388 CP4784 5188-5314 5188-5315
CA0100-1332PK 126.42 CAD
CA0100-1332 CA5062-3580 CA5062-3581 CA5062-3538 CA5181-3308 CA76508-056 CA76508-092 CA76514-660 CA76514-662 CAAG500-1004 CAAGCR213200 CAAG5181-3388 CAAGCP4784 AG5188-5314 AG5188-5315
CrossLab Graphite/Vespel Ferrules for GC
Fittings and Connectors
Graphite/Vespel (15/85%) ferrules, available in different configurations, provide leak-free connections between the column and injector on your GC.
  • Composed of 85% Vespel polyimide and 15% graphite
  • Non-porous material makes them ideal for MS applications
  • Withstand temperatures as high as 350 °C
  • Can be reused and reinstalled due their resistance to deformation
These ferrules are ideal for MS applications as their nonporous material prohibits oxygen contamination, and they can withstand temperatures as high as 350 °C. They are also widely used in GCMore Product Information