CrossLab Gas Chromatography (GC) Interface Supplies for ICP-MS
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
0100-1331 G3158-80074 G3158-60010 G3158-80060 G3158-80080 G3158-80081 G3158-80150 0100-1326 0100-1328 0100-1333 0100-1372 0100-1379 0100-1381 0100-1383 0100-1389 UCU321X UCU401X VLZF5V10 A6358 C-FP020
CA0100-1331PK 128.93 CAD
CA0100-1331 CAAGG315880074 CAAGG315860010 CAAGG315880060 CAAGG315880080 CAAGG315880081 CAAGG315880150 AG0100-1326 AG0100-1328 AG0100-1333 AG0100-1372 AG0100-1379 AG0100-1381 AG0100-1383 AG0100-1389 AGUCU321X AGUCU401X AGVLZF5V10 AGA6358 AGC-FP020
CrossLab Gas Chromatography (GC) Interface Supplies for ICP-MS
Chromatography Supplies Chromatography Spare Parts
Agilent is an expert in speciation analysis, offering GC-ICP-MS parts and interface supplies.
  • The Agilent GC-ICP-MS interface consists of a heated, passivated transfer line and a special torch with a heated injector tube
  • The unique interface is ideal for routine analysis of high-boiling point compounds, providing unparalleled performance
  • All GC-ICP-MS interface supplies are designed to Agilent’s demanding specifications and can help to achieve the market-leading ICP-MS performance that our valued clients have come to expect
  • The GC interface function is
More Product Information