Anti-SorLA/SORL1 Rabbit Monoclonal Antibody [clone: EPR23262-4]

Supplier: Abcam
AB302509-1MG AB302509-100UG
ABCA_AB302509-1MGEA 16158.66 CAD
ABCA_AB302509-1MG ABCA_AB302509-100U
Anti-SorLA/SORL1 Rabbit Monoclonal Antibody [clone: EPR23262-4]
Antibodies
Rabbit monoclonal [EPR23262-4] to SorLA/SORL1 - BSA and Azide free.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences,More Product Information
New Product
Order Now

SPECIFICATIONS

Antigen symbol SorLA/SORL1
Antigen name C11orf32
Environmentally Preferable
Antigen synonyms SORL1|Sortilin-related receptor|Low-density lipoprotein receptor relative with 11 ligand-binding repeats|SorLA-1|Sorting protein-related receptor containing LDLR class A repeats|LDLR relative with 11 ligand-binding repeats|LR11|SorLA
Antibody type Primary
Clonality Monoclonal
Clone EPR23262-4
Reactivity
Host Rabbit
Isotype IgG
ImmunoChemistry Yes
ImmunoFluorescence Yes
Immunogen The exact immunogen used to generate this antibody is proprietary information.
Form Liquid
Molecular weight 248426 Da
Purification Protein A Affinity purification
Storage temperature Store at +4 °C.
Shipping temperature Shipped at +4 °C
Tested applications ICC/IF, IHC-Fr

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR