Aloin

Supplier: Thermo Scientific Chemicals
Warning

J62153-03 J62153-MC J62153-MD
AAJ62153-03EA 420.28 CAD
AAJ62153-03 AAJ62153-MC AAJ62153-MD
Aloin
Aloin

Aloe exudant, used as a stimulant-laxative


CAS: 1415-73-2 Form: Powder Color: Yellow to brown MDL Number: MFCD00151160 Molecular Formula: C21H22O9 Formula weight: 418.40 Synonyms: Barbaloin, 1,8-Dihydroxy-10-(beta-D-glucopyranosyl)-3-(hydroxymethyl)-9(10H)-anthracenone Savety Data Phrase: Warning Beilstein Reference: 6077558 GHS H Statement:H315-H319-H335 GHS P Statement: P261-P280-P305+P351+P338-P304+P340-P405-P501a

Formula: C₂₁H₂₂O₉
Storage Temperature: Ambient
MDL Number: MFCD00151160
CAS Number: 1415-73-2
EINECS: 215-808-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR