4-Iodo-2-methoxynicotinaldehyde 95%

Supplier: Thermo Scientific Chemicals
Warning

H66984-03 H66984-MD
AAH66984-03EA 282.82 CAD
AAH66984-03 AAH66984-MD
4-Iodo-2-methoxynicotinaldehyde 95%
4-Iodo-2-methoxynicotinaldehyde

Purity: 95% CAS No.: 158669-26-2 Molecular Formula: C7H6INO2 Formula Weight: 263.03 Storage Temperature: -30°C to -10°C Physical Form: Solid Sensitivity: Light Sensitive Beilstein Reference: 7020882 MDL No.: MFCD05662396

Formula: C₇H₆INO₂
Storage Temperature: Freezer
MDL Number: MFCD05662396
CAS Number: 158669-26-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR