IEICO, OPV - Organic Photovoltaics

Supplier: Thermo Scientific Chemicals

H66656-MC H66656-MD H66656-ME
AAH66656-MCEA 861.1 CAD
AAH66656-MC AAH66656-MD AAH66656-ME
IEICO, OPV - Organic Photovoltaics
IEICO
Formula: C₁₁₄H₁₁₈N₄O₄S₄
MW: 1736.44 g/mol
CAS Number: 2055812-53-6

Order Now

Specification Test Results

Appearance (Color) Black
Form Flake or threadlike
Bandgap energy 1.34 ± 0.02 eV


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR