PBDB-T (PCE12), OPV - Organic Photovoltaics

Supplier: Thermo Scientific Chemicals

H66526-MC H66526-MD H66526-ME
AAH66526-MCEA 671.5 CAD
AAH66526-MC AAH66526-MD AAH66526-ME
PBDB-T (PCE12), OPV - Organic Photovoltaics
PBDB-T (PCE12)
Formula: (C₆₈H₇₈O₂S₈)n
CAS Number: 1415929-80-4

Order Now

Specification Test Results

Appearance (Color) Black
Form Flake or threadlike
Bandgap energy 1.82 ± 0.02 eV


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR