PPDT2FBT (PCE9.3), Organic Photovoltaics (OPV) reagent

Supplier: Thermo Scientific Chemicals

H66399-MC H66399-MD H66399-ME
AAH66399-MCEA 671.5 CAD
AAH66399-MC AAH66399-MD AAH66399-ME
PPDT2FBT (PCE9.3), Organic Photovoltaics (OPV) reagent
PPDT2FBT (PCE9.3)
Formula: (C₅₂H₇₂F₂N₂O₂S₃)n
CAS Number: 1620673-07-5

Order Now

Specification Test Results

Appearance (Color) Black
Form Flake or threadlike
Bandgap energy 1.78 ± 0.02 eV


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR