2-Bromo-5-(hydroxymethyl)phenol 95%

Supplier: Thermo Scientific Chemicals
Warning

H66253-03 H66253-MD
AAH66253-03EA 347.6 CAD
AAH66253-03 AAH66253-MD
2-Bromo-5-(hydroxymethyl)phenol 95%
2-Bromo-5-(hydroxymethyl)phenol

Purity: 95% CAS No.: 2737-19-1 Molecular Formula: C7H7BrO2 Formula Weight: 203.04 Physical Form: Solid

Formula: C₇H₇BrO₂
Storage Temperature: Ambient
MDL Number: MFCD14705103
CAS Number: 2737-19-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR