7-Methoxy-2,2,4-trimethyl-1,2-dihydroquinoline 95%

Supplier: Thermo Scientific Chemicals
Warning

H66188-03 H66188-06 H66188-MD
AAH66188-03EA 102.07 CAD
AAH66188-03 AAH66188-06 AAH66188-MD
7-Methoxy-2,2,4-trimethyl-1,2-dihydroquinoline 95%
7-Methoxy-2,2,4-trimethyl-1,2-dihydroquinoline

Purity: 95% CAS No.: 1810-74-8 Molecular Formula: C13H17NO Formula Weight: 203.29 Physical Form: Solid

Formula: C₁₃H₁₇NO
Storage Temperature: Ambient
MDL Number: MFCD00627122
CAS Number: 1810-74-8

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR