PffBT4T-2OD (PCE11), OPV - Organic Photovoltaics

Supplier: Thermo Scientific Chemicals

H66126-MC H66126-MD H66126-ME
AAH66126-MCEA 922.72 CAD
AAH66126-MC AAH66126-MD AAH66126-ME
PffBT4T-2OD (PCE11), OPV - Organic Photovoltaics
PffBT4T-2OD (PCE11)
Formula: (C₆₂H₈₈N₂S₅F₂)n
CAS Number: 1644164-62-4

Order Now

Specification Test Results

Appearance (Color) Black
Form Flake or threadlike
Bandgap energy 1.63 ± 0.02 eV


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR