PffBT4T-2DT, OPV - Organic Photovoltaics

Supplier: Thermo Scientific Chemicals

H66014-MC H66014-MD H66014-ME
AAH66014-MCEA 921.14 CAD
AAH66014-MC AAH66014-MD AAH66014-ME
PffBT4T-2DT, OPV - Organic Photovoltaics
PffBT4T-2DT
Formula: (C₆₆H₉₆N₂S₅F₂)n
CAS Number: 1430201-60-7

Order Now

Specification Test Results

Appearance (Color) Black
Form Flake or threadlike
Bandgap energy 1.65 ± 0.02 eV


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR