3-Bromo-2-fluoro-5-(trifluoromethyl)pyridine 97%

Supplier: Thermo Scientific Chemicals
Danger

H64748-03 H64748-06 H64748-MD
AAH64748-03EA 144.25 CAD
AAH64748-03 AAH64748-06 AAH64748-MD
3-Bromo-2-fluoro-5-(trifluoromethyl)pyridine 97%
3-Bromo-2-fluoro-5-(trifluoromethyl)pyridine

3-Bromo-2-fluoro-5-(trifluoromethyl)pyridine, 97%


Purity: 97% CAS No.: 1031929-01-7 Molecular Formula: C6H2BrF4N Formula Weight: 243.99 Flash Point: 30°(86°F) Physical Form: Liquid Density: 1.774 Refractive Index: 1.4600 UN No.: 1992 MDL No.: MFCD08061304

Formula: C₆H₂BrF₄N
MW: 243.987 g/mol
Density: 1.774
Flash Pt: 30 °C (86 °F)
Storage Temperature: Ambient
MDL Number: MFCD08061304
CAS Number: 1031929-01-7
UN: 1992
ADR: 3 III

Order Now

Specification Test Results

Assay 97%

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR