Isopropyl benzenesulfonate 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Benzenesulphonic acid isopropyl ester, Benzenesulfonic acid isopropyl ester

H64523-03 H64523-06 H64523-14
AAH64523-03EA 73.63 CAD
AAH64523-03 AAH64523-06 AAH64523-14
Isopropyl benzenesulfonate 95%
Isopropyl benzenesulfonate

Benzenesulfonic acid isopropyl ester, 95%


Purity: 95% CAS No.: 6214-18-2 Molecular Formula: C9H12O3S Formula Weight: 200.25 Storage Temperature: 2°C to 8°C Physical Form: Liquid Refractive Index: 1.5020 Appearance: Colorless to pale yellow Beilstein Reference: 1106314 MDL No.: MFCD04038771

Formula: C₉H₁₂O₃S
Storage Temperature: Ambient
MDL Number: MFCD04038771
CAS Number: 6214-18-2

Order Now

Specification Test Results

Assay 95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR