(R)-Methyl-2-amino-3-hydroxypropanoate hydrochloride 95%

Supplier: Thermo Scientific Chemicals
Danger

H63685-06 H63685-14 H63685-22
AAH63685-06EA 44.24 CAD
AAH63685-06 AAH63685-14 AAH63685-22
(R)-Methyl-2-amino-3-hydroxypropanoate hydrochloride 95%
(R)-Methyl-2-amino-3-hydroxypropanoate hydrochloride

D-Serine methyl ester hydrochloride, 95%


Purity: 95% CAS No.: 5874-57-7 Molecular Formula: C4H9NO3.HCl Formula Weight: 155.58 Physical Form: Solid Melting Point: 163-166° dec Optical Rotation: -4°(c=4 in ethanol) MDL No.: MFCD00066121

Formula: C4H10ClNO3
Melting Pt: 163…166 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00066121
CAS Number: 5874-57-7

Order Now

Specification Test Results

Assay 95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR