5-Methoxy-7-methyl-1H-indole 97%

Supplier: Thermo Scientific Chemicals
Warning

H63123-03 H63123-06
AAH63123-03EA 295.46 CAD
AAH63123-03 AAH63123-06 AAH63123-MD
5-Methoxy-7-methyl-1H-indole 97%
5-Methoxy-7-methyl-1H-indole

5-Methoxy-7-methylindole, 97%


Purity: 97% CAS No.: 61019-05-4 Molecular Formula: C10H11NO Formula Weight: 161.2 Physical Form: Solid MDL No.: MFCD09027022

Formula: C₁₀H₁₁NO
Storage Temperature: Ambient
MDL Number: MFCD09027022
CAS Number: 61019-05-4

Order Now

Specification Test Results

Assay 97%

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR