Ethyl-4,6-dichloronicotinate 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ethyl 4,6-dichloronicotinate

H63092-03 H63092-06
AAH63092-03EA 109.49 CAD
AAH63092-03 AAH63092-06 AAH63092-MD
Ethyl-4,6-dichloronicotinate 95%
Ethyl-4,6-dichloronicotinate

Ethyl 4,6-dichloronicotinate, 95%


Purity: 95% CAS No.: 40296-46-6 Molecular Formula: C8H7Cl2NO2 Formula Weight: 220.05 Storage Temperature: -30°C to -10°C Physical Form: Liquid Appearance: Yellow MDL No.: MFCD00234408

Formula: C8H7Cl2NO2
MW: 220.05 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00234408
CAS Number: 40296-46-6

Order Now

Specification Test Results

Assay 95%

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR