D-(-)-Cysteine 98%

Supplier: Thermo Scientific Chemicals

H62627-03 H62627-06
AAH62627-03EA 127.35 CAD
AAH62627-03 AAH62627-06
D-(-)-Cysteine 98%
D(-)-Cysteine

D-Cysteine, 98%


Purity: 98% CAS No.: 921-01-7 Molecular Formula: C3H7NO2S Formula Weight: 121.16 Physical Form: Solid Sensitivity: Air Sensitive Melting Point: 230° dec Beilstein Reference: 1721407 MDL No.: MFCD00066461

Formula: C₃H₇NO₂S
Melting Pt: 230 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00066461
CAS Number: 921-01-7
EINECS: 213-062-0

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR