(S)-4-Amino-2-(tert-butoxycarbonylamino)butyric acid 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Boc-Dab-OH, (S)-4-Amino-2-(Boc-amino)butyric Acid, (S)-4-Amino-2-(tert-butoxycarbonylamino)butyric Acid, (S)-Nalpha-Boc-2,4-diaminobutyric Acid, (S)-4-Amino-2-(Boc-amino)butyric acid (S)-Nα-Boc-2,4-diaminobutyric acid Boc-Dab-OH

H62450-03 H62450-06
AAH62450-03EA 78.37 CAD
AAH62450-03 AAH62450-06 AAH62450-MD
(S)-4-Amino-2-(tert-butoxycarbonylamino)butyric acid 97%
(S)-4-Amino-2-(tert-butoxycarbonylamino)butyric acid

(S)-4-Amino-2-(Boc-amino)butyric acid, 97%


Purity: 97% CAS No.: 25691-37-6 Molecular Formula: C9H18N2O4 Formula Weight: 218.25 Physical Form: Powder Appearance: White to off-white Beilstein Reference: 4745487 Optical Rotation: +7°(c=1 in methanol) MDL No.: MFCD00236841

Formula: C₉H₁₈N₂O₄
MW: 218.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00236841
CAS Number: 25691-37-6

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR