Fmoc-D-Dap(Boc)-OH 95%

Supplier: Thermo Scientific Chemicals

H62381-03 H62381-06
AAH62381-03EA 222.78 CAD
AAH62381-03 AAH62381-06 AAH62381-MD
Fmoc-D-Dap(Boc)-OH 95%
Fmoc-D-Dap(Boc)-OH

(R)-3-(Boc-amino)-2-(Fmoc-amino)propionic acid, 95%


Purity: 95% CAS No.: 198544-42-2 Molecular Formula: C23H26N2O6 Formula Weight: 426.46 Physical Form: Solid MDL No.: MFCD00798648

Formula: C₂₃H₂₆N₂O₆
Storage Temperature: Ambient
MDL Number: MFCD00798648
CAS Number: 198544-42-2

Order Now

Specification Test Results

Assay 95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR