5-Bromopyrimidine-2-carbonitrile 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Bromo-2-cyanopyrimidine, 5-bromo-pyrimidine-2-carbonitile

H61363-03 H61363-06
AAH61363-03EA 69.36 CAD
AAH61363-03 AAH61363-06
5-Bromopyrimidine-2-carbonitrile 95%
5-Bromopyrimidine-2-carbonitrile

5-Bromo-2-cyanopyrimidine, 95%


Purity: 95% CAS No.: 38275-57-9 Molecular Formula: C5H2BrN3 Formula Weight: 183.99 Storage Temperature: 2°C to 8°C Physical Form: Solid Sensitivity: Moisture Sensitive Appearance: White to yellow Beilstein Reference: 775324 UN No.: 3439 MDL No.: MFCD02940446

Formula: C₅H₂BrN₃
MW: 183.99 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02940446
CAS Number: 38275-57-9
UN: 3439
ADR: 6.1 III

Order Now

Specification Test Results

Assay 95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR