1-(4-Bromo-3-fluorophenyl)cyclopropanecarbonitrile 96%

Supplier: Thermo Scientific Chemicals
Warning

H59323-03 H59323-MD
AAH59323-03EA 636.74 CAD
AAH59323-03 AAH59323-MD
1-(4-Bromo-3-fluorophenyl)cyclopropanecarbonitrile 96%
1-(4-Bromo-3-fluorophenyl)cyclopropanecarbonitrile

Purity: 96% CAS No.: 749269-73-6 Molecular Formula: C10H7BrFN Formula Weight: 240.07 UN No.: 3439 MDL No.: MFCD19692057

Formula: C₁₀H₇BrFN
Storage Temperature: Ambient
MDL Number: MFCD19692057
CAS Number: 749269-73-6
UN: 3439
ADR: "6.1",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR