Pyrrole-3-carboxylic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning

H55383-06
AAH55383-06EA 497.7 CAD
AAH55383-06
Pyrrole-3-carboxylic acid 98+%
Pyrrole-3-carboxylic acid

Purity: 98+% CAS No.: 931-03-3 Molecular Formula: C5H5NO2 Formula Weight: 111.10 Beilstein Reference: 108642 MDL No.: MFCD00800594

Formula: C₅H₅NO₂
Storage Temperature: Ambient
MDL Number: MFCD00800594
CAS Number: 931-03-3

Order Now

Specification Test Results

Assay 98%

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR