3-bromo-4-methoxybenzenesulfonyl chloride 97%

Supplier: Thermo Scientific Chemicals
Danger

H54015-06
AAH54015-03EA 103.4 CAD
AAH54015-03 AAH54015-06 AAH54015-MD
3-bromo-4-methoxybenzenesulfonyl chloride 97%
3-bromo-4-methoxybenzenesulfonyl chloride

Purity: 97% CAS No.: 23094-96-4 Molecular Formula: C7H6BrClO3S Formula Weight: 285.54 UN No.: 3265 MDL No.: MFCD03989643

Formula: C₇H₆BrClO₃S
Storage Temperature: Ambient
MDL Number: MFCD03989643
CAS Number: 23094-96-4
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR