Pyridine, anhydrous ≥99.5%, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger


Packaged under argon.

047323-AE 047323-K2
AA47323-AEEA 289.05 CAD
AA47323-AE AA47323-K2
Pyridine, anhydrous ≥99.5%, AcroSeal™
Pyridine

AcroSeal™ septum bottled solvent


  • Supports optimum performance of your moisture-sensitive work

Formula: C₅H₅N
Boiling Pt: 115 °C
Melting Pt: –42 °C
Density: 0.978
Flash Pt: 20 °C (68 °F)
Storage Temperature: Ambient
MDL Number: MFCD00011732
CAS Number: 110-86-1
EINECS: 203-809-9
UN: 1282
ADR: 3,
Merck Index: 14,7970
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3f9c5934940218027dae0d80a1f98b6d5179c0a8&vid=9f2ef46a26e488d366e82869b3acb96652b5dab1&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR